siRNA Details
Gene Symbol: Pde6c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170959.1 | NP_001164430.1 | 10 | 1479 | ||
NM_033614.2 | NP_291092.1 | 10 | 1479 | |||
GenBank mRNA | AF411063.1 | - | 10 | 1314 | ||
BC027050.1 | - | 10 | 1479 | 5358611 |
