siRNA Details
| Gene Symbol: Pde6c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170959.1 | NP_001164430.1 | 5 | 1066 | ||
| NM_033614.2 | NP_291092.1 | 5 | 1066 | |||
| GenBank mRNA | AF411063.1 | - | 5 | 901 | ||
| BC027050.1 | - | 5 | 1066 | 5358611 | ||

Pde6c
Genomic Map