siRNA Details
| Gene Symbol: Slc8a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_148946.2 | NP_683748.1 | 8 | 2210 | ||
| GenBank mRNA | AF503502.1 | - | 7 | 2075 | ||
| AK035163.1 | - | 8 | 2352 | |||
| BC058704.1 | - | 8 | 2210 | 6838206 | ||
| BC080277.1 | - | 7 | 2144 | 30354551 | ||

Slc8a2
Genomic Map