siRNA Details
| Gene Symbol: Cds2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291039.1 | NP_001277968.1 | 5 | 478 | ||
| NM_138651.7 | NP_619592.1 | 5 | 801 | |||
| GenBank mRNA | AK036328.1 | - | 5 | 652 | ||
| AK049257.1 | - | 5 | 479 | |||
| AK147541.1 | - | 5 | 693 | |||
| AK166001.1 | - | 5 | 652 | |||
| AY159802.1 | - | 5 | 724 | |||
| BC003852.1 | - | 5 | 707 | 3600825 | ||
| BC069879.1 | - | 5 | 680 | 30439069 | ||
| CD540005.1 | - | 5 | 476 | |||

Cds2
Genomic Map