siRNA Details
| Gene Symbol: Cds2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291039.1 | NP_001277968.1 | 6 | 516 | ||
| NM_138651.7 | NP_619592.1 | 6 | 839 | |||
| GenBank mRNA | AK036328.1 | - | 6 | 690 | ||
| AK049257.1 | - | 6 | 517 | |||
| AK147541.1 | - | 6 | 731 | |||
| AK166001.1 | - | 6 | 690 | |||
| AY159802.1 | - | 6 | 762 | |||
| BC003852.1 | - | 6 | 745 | 3600825 | ||
| BC069879.1 | - | 6 | 718 | 30439069 | ||
| CD540005.1 | - | 6 | 514 | |||

Cds2
Genomic Map