siRNA Details
| Gene Symbol: Atp6v1b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134157.2 | NP_598918.1 | 10 | 1115 | ||
| GenBank mRNA | AF435091.1 | - | 10 | 1115 | ||
| AK052604.1 | - | 10 | 1086 | |||
| AK052707.1 | - | 10 | 1077 | |||
| AK078810.1 | - | 10 | 1143 | |||
| AK085549.1 | - | 10 | 1116 | |||
| AK168980.1 | - | 10 | 1143 | |||
| BC004789.1 | - | 7 | 709 | |||
| BC017127.1 | - | 10 | 1100 | 4489144 | ||
| BC062202.1 | - | 10 | 1074 | 2650839 | ||

Atp6v1b1
Genomic Map