siRNA Details
Gene Symbol: Hlcs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139145.4 | NP_631884.1 | 4 | 1213 | ||
GenBank mRNA | AB066227.1 | - | 3 | 1073 | ||
AK052424.1 | - | 5 | 1509 | |||
AK158212.1 | - | 5 | 1330 | |||
AK164351.1 | - | 5 | 1494 | |||
BC050090.1 | - | 4 | 1214 | 30012868 |
