siRNA Details
| Gene Symbol: M1ap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033079.2 | NP_149070.2 | 8 | 1348 | ||
| GenBank mRNA | AF084364.1 | - | 7 | 1317 | ||
| AK131954.1 | - | 8 | 1348 | |||
| BC119803.1 | - | 7 | 1275 | 40107692 | ||
| BC122521.1 | - | 7 | 1280 | 40107688 | ||
| BQ553015.1 | - | 2 | 559 | |||

M1ap
Genomic Map