siRNA Details
| Gene Symbol: C030039L03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001112731.1 | NP_001106202.1 | 6 | 678 | ||
| NM_198417.2 | NP_940809.2 | 6 | 658 | |||
| GenBank mRNA | AK140966.1 | - | 6 | 582 | ||
| AK141645.1 | - | 6 | 579 | |||
| BC060708.1 | - | 6 | 658 | 6406744 | ||
| BQ770435.1 | - | 1 | 110 | |||

C030039L03Rik
Genomic Map