siRNA Details
| Gene Symbol: 2010002M12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053217.2 | NP_444447.1 | 3 | 784 | ||
| GenBank mRNA | AK008052.1 | - | 2 | 636 | ||
| AK136525.1 | - | 2 | 636 | |||
| AK153247.1 | - | 3 | 785 | |||
| BC003493.1 | - | 1 | 139 | |||
| BC138202.1 | - | 2 | 615 | 8861221 | ||

2010002M12Rik
Genomic Map