siRNA Details
| Gene Symbol: Pzp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007376.4 | NP_031402.3 | 26 | 3344 | ||
| GenBank mRNA | BC057983.1 | - | 26 | 3344 | 5134629 | |
| M93264.1 | - | 26 | 3337 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pzp| Gene Symbol: Pzp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007376.4 | NP_031402.3 | 26 | 3344 | ||
| GenBank mRNA | BC057983.1 | - | 26 | 3344 | 5134629 | |
| M93264.1 | - | 26 | 3337 | |||