siRNA Details
Gene Symbol: Abca1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013454.3 | NP_038482.3 | 28, 29 | 4359 | ||
GenBank mRNA | AK051920.1 | - | 28, 29 | 4360 | ||
X75926.1 | - | 27, 28 | 4122 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Abca1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013454.3 | NP_038482.3 | 28, 29 | 4359 | ||
GenBank mRNA | AK051920.1 | - | 28, 29 | 4360 | ||
X75926.1 | - | 27, 28 | 4122 |