siRNA Details
| Gene Symbol: Abca4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007378.1 | NP_031404.1 | 31, 32 | 4709 | ||
| GenBank mRNA | AF000149.1 | - | 31, 32 | 4709 | ||
| AK052916.1 | - | 8, 9 | 1067 | |||
| BC057853.1 | - | 31, 32 | 4709 | 6492139 | ||

Abca4
Genomic Map