siRNA Details
| Gene Symbol: Abcg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009593.2 | NP_033723.1 | 3, 4 | 527 | ||
| GenBank mRNA | AF323659.1 | - | 3, 4 | 523 | ||
| AK037433.1 | - | 3, 4 | 447 | |||
| BC119471.2 | - | 3, 4 | 475 | 40048166 | ||
| U34920.1 | - | 3, 4 | 485 | |||
| Z48745.1 | - | 3, 4 | 514 | |||

Abcg1
Genomic Map