siRNA Details
| Gene Symbol: Abcg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009593.2 | NP_033723.1 | 14 | 1820 | ||
| GenBank mRNA | AF323659.1 | - | 14 | 1816 | ||
| BC119471.2 | - | 14 | 1768 | 40048166 | ||
| CO431055.1 | - | 2 | 65 | |||
| CV558133.1 | - | 6 | 631 | |||
| U34920.1 | - | 14 | 1778 | |||
| Z48745.1 | - | 14 | 1807 | |||

Abcg1
Genomic Map