siRNA Details
| Gene Symbol: Acadm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007382.5 | NP_031408.1 | 11, 12 | 1404 | ||
| GenBank mRNA | AK079930.1 | - | 3, 4 | 448 | ||
| AK146444.1 | - | 11, 12 | 1209 | |||
| BC013498.1 | - | 11, 12 | 1189 | 4222450 | ||
| U07159.1 | - | 11, 12 | 1193 | |||

Acadm
Genomic Map