siRNA Details
| Gene Symbol: Acadvl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017366.3 | NP_059062.1 | 8 | 742 | ||
| GenBank mRNA | AF017176.1 | - | 5 | 390 | ||
| AK076037.1 | - | 8 | 703 | |||
| AK146334.1 | - | 3 | 215 | |||
| AK196376.1 | - | 3 | 305 | |||
| AK215457.1 | - | 3 | 305 | |||
| BC026559.1 | - | 8 | 721 | 5037878 | ||
| Z71189.1 | - | 8 | 663 | |||

Acadvl
Genomic Map