siRNA Details
| Gene Symbol: Vmn1r43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053220.2 | NP_444450.2 | 1 | 739 | ||
| GenBank mRNA | BC125364.1 | - | 1 | 739 | 40129879 | |
| BC132284.1 | - | 1 | 739 | 40130561 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r43| Gene Symbol: Vmn1r43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053220.2 | NP_444450.2 | 1 | 739 | ||
| GenBank mRNA | BC125364.1 | - | 1 | 739 | 40129879 | |
| BC132284.1 | - | 1 | 739 | 40130561 | ||