siRNA Details
| Gene Symbol: Laptm4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033521.3 | NP_277056.1 | 4 | 603 | ||
| GenBank mRNA | AF317418.1 | - | 4 | 305 | ||
| AK077269.1 | - | 4 | 466 | |||
| AK077933.1 | - | 4 | 603 | |||
| AK079581.1 | - | 4 | 466 | |||
| AK082807.1 | - | 4 | 482 | |||
| AK146868.1 | - | 3 | 369 | |||
| AK166014.1 | - | 4 | 505 | |||
| BC019120.1 | - | 4 | 473 | 4507607 | ||
| EU561010.1 | - | 4 | 603 | |||

Laptm4b
Genomic Map