siRNA Details
| Gene Symbol: Cldn16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053241.5 | NP_444471.1 | 4 | 912 | ||
| GenBank mRNA | AB009016.1 | - | 4 | 574 | ||
| AF323748.1 | - | 4 | 702 | |||
| AK085268.1 | - | 4 | 913 | |||
| AK085333.1 | - | 4 | 888 | |||
| BC115574.2 | - | 5 | 691 | 40102178 | ||
| BC115575.2 | - | 5 | 691 | 40102179 | ||

Cldn16
Genomic Map