siRNA Details
| Gene Symbol: Foxp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286607.1 | NP_001273536.1 | 12 | 1782 | ||
| NM_053242.4 | NP_444472.2 | 14 | 2182 | |||
| NM_212435.1 | NP_997600.1 | 12 | 1845 | |||
| GenBank mRNA | AF339106.1 | - | 11 | 1523 | ||
| AK051668.1 | - | 14 | 2165 | |||
| AK082361.1 | - | 14 | 1854 | |||
| AY079003.1 | - | 14 | 1811 | |||
| BC058960.1 | - | 12 | 1779 | 6811877 | ||
| BC062926.1 | - | 12 | 1845 | 6851153 | ||

Foxp2
Genomic Map