siRNA Details
Gene Symbol: Slc33a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272035.1 | NP_001258964.1 | 3 | 1576 | ||
NM_015728.5 | NP_056543.2 | 3 | 1576 | |||
GenBank mRNA | AB016795.1 | - | 3 | 1033 | ||
AK144703.1 | - | 3 | 1443 | |||
AW519598.1 | - | 1 | 55 | |||
BC005616.1 | - | 3 | 1153 | 3154886 | ||
BC054395.1 | - | 3 | 1576 | 6492236 | ||
BC063104.1 | - | 1509 | 5042025 |
