siRNA Details
| Gene Symbol: Palmd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023245.3 | NP_075734.3 | 7 | 1308 | ||
| GenBank mRNA | AF263246.1 | - | 7 | 1012 | ||
| AJ312215.1 | - | 7 | 1031 | |||
| AK014531.1 | - | 7 | 1106 | |||
| AK136939.1 | - | 7 | 1215 | |||
| AK171869.1 | - | 7 | 1039 | |||
| AL391037.1 | - | 7 | 1012 | |||
| BC010193.1 | - | 7 | 1072 | 3156945 | ||

Palmd
Genomic Map