siRNA Details
| Gene Symbol: Acox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271898.1 | NP_001258827.1 | 9 | 1557 | ||
| NM_015729.3 | NP_056544.2 | 9 | 1557 | |||
| GenBank mRNA | AB034914.1 | - | 9 | 1246 | ||
| AF006688.1 | - | 9 | 1296 | |||
| AK034760.1 | - | 5 | 862 | |||
| AK040566.1 | - | 9 | 1260 | |||
| AK054446.1 | - | 9 | 1333 | |||
| AK128941.1 | - | 9 | 1259 | |||
| AK170217.1 | - | 5 | 627 | |||
| BC054727.1 | - | 9 | 1225 | |||
| BC056448.1 | - | 9 | 1557 | 5704873 | ||

Acox1
Genomic Map