siRNA Details
| Gene Symbol: Chrna4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015730.5 | NP_056545.3 | 5 | 1410 | ||
| GenBank mRNA | AF225912.2 | - | 5 | 1308 | ||
| AK034228.1 | - | 5 | 1421 | |||
| AK083157.1 | - | 5 | 1341 | |||
| AK136886.1 | - | 5 | 1397 | |||
| AK143938.1 | - | 5 | 1418 | |||
| AY574263.1 | - | 5 | 1306 | |||
| BC053013.1 | - | 5 | 1410 | 5706867 | ||

Chrna4
Genomic Map