siRNA Details
| Gene Symbol: Chrna6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021369.2 | NP_067344.2 | 5 | 1000 | ||
| GenBank mRNA | AJ245706.1 | - | 5 | 838 | ||
| AK149262.1 | - | 5 | 941 | |||
| AY574265.1 | - | 5 | 758 | |||
| BC031985.1 | - | 5 | 1000 | 4501558 | ||

Chrna6
Genomic Map