siRNA Details
| Gene Symbol: Chrnd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021600.3 | NP_067611.2 | 8, 9 | 1032 | ||
| GenBank mRNA | AK081344.1 | - | 7, 8 | 1018 | ||
| BC052153.1 | - | 8, 9 | 977 | 6336578 | ||
| BC141376.1 | - | 8, 9 | 960 | 9055989 | ||
| BC141387.1 | - | 8, 9 | 960 | 9056000 | ||
| K02582.1 | - | 8, 9 | 943 | |||
| L10076.1 | - | 8, 9 | 992 | |||

Chrnd
Genomic Map