siRNA Details
Gene Symbol: Chrne | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009603.1 | NP_033733.1 | 4 | 265 | ||
GenBank mRNA | AF032863.1 | - | 3 | 118 | ||
X55718.1 | - | 4 | 265 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Chrne | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009603.1 | NP_033733.1 | 4 | 265 | ||
GenBank mRNA | AF032863.1 | - | 3 | 118 | ||
X55718.1 | - | 4 | 265 |