siRNA Details
| Gene Symbol: Actn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013456.2 | NP_038484.1 | 21 | 2744 | ||
| GenBank mRNA | AF093775.1 | - | 21 | 2719 | ||
| AK134757.1 | - | 6 | 848 | |||
| BC111890.1 | - | 21 | 2710 | 40060598 | ||
| BX630701.1 | - | 1 | 143 | |||

Actn3
Genomic Map