siRNA Details
| Gene Symbol: Acvr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110204.1 | NP_001103674.1 | 11 | 1850 | ||
| NM_001110205.1 | NP_001103675.1 | 9 | 1328 | |||
| NM_007394.3 | NP_031420.2 | 10 | 1605 | |||
| GenBank mRNA | AF332087.1 | - | 8 | 1238 | ||
| AF332088.1 | - | 8 | 1238 | |||
| AK052246.1 | - | 1440 | ||||
| AK053428.1 | - | 11 | 1849 | |||
| AK150014.1 | - | 10 | 1605 | |||
| AK150075.1 | - | 10 | 1605 | |||
| BC058718.1 | - | 8 | 1326 | 6418954 | ||
| L15436.1 | - | 9 | 1678 | |||

Acvr1
Genomic Map