siRNA Details
| Gene Symbol: Acvr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110204.1 | NP_001103674.1 | 11, 12 | 1922 | ||
| NM_001110205.1 | NP_001103675.1 | 9, 10 | 1400 | |||
| NM_007394.3 | NP_031420.2 | 10, 11 | 1677 | |||
| GenBank mRNA | AF332087.1 | - | 8, 9 | 1310 | ||
| AF332088.1 | - | 8, 9 | 1310 | |||
| AK053428.1 | - | 11, 12 | 1921 | |||
| AK150014.1 | - | 10, 11 | 1677 | |||
| AK150075.1 | - | 10, 11 | 1677 | |||
| BC058718.1 | - | 8, 9 | 1398 | 6418954 | ||
| L15436.1 | - | 9, 10 | 1750 | |||

Acvr1
Genomic Map