siRNA Details
| Gene Symbol: Acvr1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007395.3 | NP_031421.1 | 5 | 965 | ||
| GenBank mRNA | AK154578.1 | - | 988 | |||
| AK157910.1 | - | 5 | 966 | |||
| BC059832.1 | - | 958 | ||||
| BC145775.1 | - | 5 | 1018 | 40131078 | ||
| BC145777.1 | - | 5 | 1018 | 40131080 | ||
| Z31663.1 | - | 5 | 954 | |||

Acvr1b
Genomic Map