siRNA Details
| Gene Symbol: Aspa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023113.5 | NP_075602.2 | 3, 4 | 663 | ||
| GenBank mRNA | AF212998.1 | - | 3, 4 | 513 | ||
| AK002859.1 | - | 3, 4 | 661 | |||
| AK008354.1 | - | 3, 4 | 666 | |||
| BC024934.1 | - | 3, 4 | 627 | 4012726 | ||

Aspa
Genomic Map