siRNA Details
Gene Symbol: Adam10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007399.3 | NP_031425.2 | 15, 16 | 2354 | ||
GenBank mRNA | AF011379.1 | - | 15, 16 | 2175 | ||
AK036599.1 | - | 9, 10 | 1398 | |||
AK036883.1 | - | 7, 8 | 1042 | |||
AK036918.1 | - | 7, 8 | 1043 | |||
BC057346.1 | - | 15, 16 | 2315 |
