siRNA Details
| Gene Symbol: Adam10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007399.3 | NP_031425.2 | 12 | 1850 | ||
| GenBank mRNA | AF011379.1 | - | 12 | 1671 | ||
| AK036599.1 | - | 6 | 894 | |||
| AK036883.1 | - | 4 | 538 | |||
| AK036918.1 | - | 4 | 539 | |||
| AK217253.1 | - | 1 | 71 | |||
| BC057346.1 | - | 12 | 1811 | |||

Adam10
Genomic Map