siRNA Details
| Gene Symbol: Afg3l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054070.3 | NP_473411.2 | 13 | 1603 | ||
| GenBank mRNA | AF329695.1 | - | 13 | 1544 | ||
| AK159647.1 | - | 13 | 1567 | |||
| AK167964.1 | - | 13 | 1578 | |||
| AK168244.1 | - | 13 | 1577 | |||
| BC043320.1 | - | 4 | 425 | |||
| BC056978.1 | - | 13 | 1562 | 6821746 | ||
| BC060064.1 | - | 13 | 1562 | |||

Afg3l1
Genomic Map