siRNA Details
| Gene Symbol: Afg3l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054070.3 | NP_473411.2 | 5 | 449 | ||
| GenBank mRNA | AF329695.1 | - | 5 | 390 | ||
| AK159647.1 | - | 5 | 413 | |||
| AK167964.1 | - | 5 | 424 | |||
| AK168244.1 | - | 5 | 423 | |||
| BC056978.1 | - | 5 | 408 | 6821746 | ||
| BC060064.1 | - | 5 | 408 | |||

Afg3l1
Genomic Map