siRNA Details
| Gene Symbol: Adam12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007400.2 | NP_031426.2 | 14 | 1681 | ||
| GenBank mRNA | AK051780.1 | - | 4 | 495 | ||
| D50411.1 | - | 14 | 1681 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adam12| Gene Symbol: Adam12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007400.2 | NP_031426.2 | 14 | 1681 | ||
| GenBank mRNA | AK051780.1 | - | 4 | 495 | ||
| D50411.1 | - | 14 | 1681 | |||