siRNA Details
Gene Symbol: Adam8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK089086.1 | - | 17 | 1843 | ||
AK089169.1 | - | 17 | 1883 | |||
AK134118.1 | - | 7 | 1397 | |||
AK150411.1 | - | 17 | 1845 | |||
AK151535.1 | - | 9 | 1074 | |||
AK152675.1 | - | 17 | 1862 | |||
AK152769.1 | - | 17 | 1844 | |||
AK153065.1 | - | 17 | 1844 | |||
AK155772.1 | - | 17 | 1843 | |||
AK155912.1 | - | 17 | 1863 | |||
AK160041.1 | - | 17 | 1846 | |||
AK170019.1 | - | 17 | 1868 | |||
AK171022.1 | - | 15 | 2420 | |||
AK171060.1 | - | 17 | 1843 | |||
BC025584.1 | - | 17 | 1897 | 5324943 | ||
X13335.1 | - | 17 | 1842 |
