siRNA Details
| Gene Symbol: Adam9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270996.1 | NP_001257925.1 | 13 | 1508 | ||
| NM_007404.2 | NP_031430.2 | 13 | 1508 | |||
| GenBank mRNA | AK122188.1 | - | 13 | 1491 | ||
| AK148174.1 | - | 13 | 1394 | |||
| AK155582.1 | - | 13 | 1469 | |||
| AK155672.1 | - | 13 | 1401 | |||
| AK161778.1 | - | 13 | 1431 | |||
| BC047156.1 | - | 13 | 1386 | 6488707 | ||
| BC052710.1 | - | 13 | 1380 | |||
| D50412.1 | - | 2 | 92 | |||
| U06145.1 | - | 1 | 74 | |||
| U41765.1 | - | 13 | 1380 | |||

Adam9
Genomic Map