siRNA Details
| Gene Symbol: Adamts1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009621.4 | NP_033751.3 | 2 | 1236 | ||
| GenBank mRNA | AK156830.1 | - | 2 | 1235 | ||
| AK161409.1 | - | 2 | 338 | |||
| AK163622.1 | - | 2 | 1235 | |||
| BC003269.1 | - | 2 | 1221 | |||
| BC040382.1 | - | 2 | 1220 | 3156736 | ||
| BC050834.1 | - | 2 | 1220 | 6505494 | ||
| D67076.1 | - | 2 | 1220 | |||

Adamts1
Genomic Map