siRNA Details
| Gene Symbol: Adcy9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291910.1 | NP_001278839.1 | 4 | 2379 | ||
| NM_009624.3 | NP_033754.2 | 3 | 2602 | |||
| GenBank mRNA | AK156940.1 | - | 4 | 2381 | ||
| BE862471.1 | - | 3 | 183 | |||
| U30602.1 | - | 3 | 1909 | |||
| Z50190.1 | - | 2 | 1878 | |||

Adcy9
Genomic Map