siRNA Details
| Gene Symbol: Adcy9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291910.1 | NP_001278839.1 | 3 | 1208 | ||
| NM_009624.3 | NP_033754.2 | 2 | 1431 | |||
| GenBank mRNA | AK156940.1 | - | 3 | 1210 | ||
| U30602.1 | - | 2 | 738 | |||
| Z50190.1 | - | 1 | 707 | |||

Adcy9
Genomic Map