siRNA Details
| Gene Symbol: Parp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009632.2 | NP_033762.1 | 9 | 829 | ||
| GenBank mRNA | AF072521.1 | - | 9 | 804 | ||
| AJ007780.1 | - | 9 | 842 | |||
| BC052000.1 | - | 8 | 1214 | |||
| BC062150.1 | - | 9 | 829 | 6333875 | ||

Parp2
Genomic Map