siRNA Details
Gene Symbol: Adra1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284380.1 | NP_001271309.1 | 3 | 974 | ||
NM_001284381.1 | NP_001271310.1 | 4 | 1124 | |||
NM_007416.4 | NP_031442.2 | 1 | 970 | |||
GenBank mRNA | AK004891.1 | - | 4 | 1081 | ||
AK043877.1 | - | 1 | 815 | |||
AK139448.1 | - | 1 | 973 | |||
BC018330.1 | - | 3 | 947 | |||
BC037002.1 | - | 3 | 948 | |||
BC138653.1 | - | 1 | 688 | 8861673 | ||
BC138654.1 | - | 1 | 688 | 8861674 | ||
Y12738.1 | - | 1 | 1289 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
