siRNA Details
| Gene Symbol: Adsl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009634.6 | NP_033764.2 | 2 | 399 | ||
| GenBank mRNA | AK049372.1 | - | 2 | 371 | ||
| AK051111.1 | - | 2 | 371 | |||
| AK152092.1 | - | 2 | 371 | |||
| AK166019.1 | - | 2 | 367 | |||
| AK168906.1 | - | 2 | 364 | |||
| AK170216.1 | - | 2 | 364 | |||
| AK171725.1 | - | 2 | 380 | |||
| AK201011.1 | - | 1 | 61 | |||
| BC020187.1 | - | 2 | 375 | 4913637 | ||
| U20225.1 | - | 2 | 375 | |||

Adsl
Genomic Map