siRNA Details
| Gene Symbol: Gla | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013463.2 | NP_038491.2 | 7 | 1155 | ||
| GenBank mRNA | AK040081.1 | - | 7 | 1087 | ||
| AK054547.1 | - | 7 | 1107 | |||
| AK164231.1 | - | 7 | 1088 | |||
| BC009021.1 | - | 7 | 1108 | 3708950 | ||
| U34071.1 | - | 7 | 1105 | |||
| U50716.1 | - | 7 | 1093 | |||

Gla
Genomic Map