siRNA Details
| Gene Symbol: Gla | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013463.2 | NP_038491.2 | 1 | 252 | ||
| GenBank mRNA | AK040081.1 | - | 1 | 184 | ||
| AK054547.1 | - | 1 | 204 | |||
| AK145011.1 | - | 1 | 196 | |||
| AK164231.1 | - | 1 | 185 | |||
| AK166204.1 | - | 1 | 185 | |||
| AK166245.1 | - | 1 | 201 | |||
| BC009021.1 | - | 1 | 205 | 3708950 | ||
| U34071.1 | - | 1 | 202 | |||
| U50716.1 | - | 1 | 190 | |||

Gla
Genomic Map