siRNA Details
| Gene Symbol: Agtrap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301281.1 | NP_001288210.1 | 4 | 367 | ||
| NM_009642.4 | NP_033772.2 | 4 | 414 | |||
| GenBank mRNA | AF102548.1 | - | 4 | 293 | ||
| AK007383.1 | - | 4 | 351 | |||
| AK007502.1 | - | 4 | 326 | |||
| AK036598.1 | - | 4 | 343 | |||
| AK152467.1 | - | 4 | 414 | |||
| BC046820.1 | - | 4 | 400 | 6400135 | ||
| BC057196.1 | - | 4 | 319 | 5249164 | ||

Agtrap
Genomic Map