siRNA Details
| Gene Symbol: Ak1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198790.1 | NP_001185719.1 | 6 | 529 | ||
| NM_001198791.1 | NP_001185720.1 | 6 | 511 | |||
| NM_001198792.1 | NP_001185721.1 | 5 | 470 | |||
| NM_021515.3 | NP_067490.1 | 5 | 488 | |||
| GenBank mRNA | AJ010108.1 | - | 5 | 394 | ||
| AJ010109.1 | - | 5 | 484 | |||
| AK046613.1 | - | 6 | 530 | |||
| AK089270.1 | - | 6 | 512 | |||
| BC014802.1 | - | 6 | 455 | 4505142 | ||
| BC054366.1 | - | 5 | 477 | 5346232 | ||
| CF950379.1 | - | 5 | 488 | |||
| DQ486026.1 | - | 6 | 542 | |||

Ak1
Genomic Map